3-(4-Ethoxyphenyl)-2-methyl-1-propene ≥97%

Supplier: Matrix Scientific

086241-1G 086241-5G
102847-524EA 387.69 USD
102847-524 102847-528
3-(4-Ethoxyphenyl)-2-methyl-1-propene ≥97%
3-(4-Ethoxyphenyl)-2-methyl-1-propene

Hazard: Irritant

Formula: C₁₂H₁₆O
MDL Number: MFCD09801158
CAS Number: 951890-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR