4-(2-Ethoxyphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086237-1G 086237-5G
102847-500EA 580 USD
102847-500 102847-504
4-(2-Ethoxyphenyl)-1-butene ≥97%
4-(2-Ethoxyphenyl)-1-butene

Hazard: Irritant

Formula: C₁₂H₁₆O
MDL Number: MFCD09801131
CAS Number: 951889-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR