2-Bromo-3-[(2-methylthio)phenyl]-1-propene ≥97%

Supplier: Matrix Scientific

086231-1G 086231-5G
102847-464EA 592.31 USD
102847-464 102847-468
2-Bromo-3-[(2-methylthio)phenyl]-1-propene ≥97%
2-Bromo-3-[(2-methylthio)phenyl]-1-propene

Hazard: Irritant

Formula: C₁₀H₁₁BrS
MDL Number: MFCD07775129
CAS Number: 951889-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR