5-(3-Chloro-5-fluorophenyl)-1-pentene ≥97%

Supplier: Matrix Scientific

086201-1G 086201-2G 086201-5G
102847-284EA 790.77 USD
102847-284 102847-286 102847-288
5-(3-Chloro-5-fluorophenyl)-1-pentene ≥97%
5-(3-Chloro-5-fluorophenyl)-1-pentene

Hazard: Irritant

Formula: C₁₁H₁₂ClF
MDL Number: MFCD11915887
CAS Number: 1143461-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR