3-(4-Cyano-3-fluorophenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086199-1G 086199-2G 086199-5G
102847-272EA 387.69 USD
102847-272 102847-274 102847-276
3-(4-Cyano-3-fluorophenyl)-1-propene ≥97%
3-(4-Cyano-3-fluorophenyl)-1-propene

Hazard: Irritant

Formula: C₁₀H₈FN
MDL Number: MFCD09801087
CAS Number: 951888-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR