4-Hydroxy-6-methyl-3,4-dihydro-2H-pyran-2-one ≥97%
Supplier: Matrix Scientific
085485-1G
085485-10G
102845-156EA
72.31
USD
102845-156
102845-158
4-Hydroxy-6-methyl-3,4-dihydro-2H-pyran-2-one ≥97%
4-Hydroxy-6-methyl-3,4-dihydro-2H-pyran-2-one
Hazard: Irritant
Formula:
C₆H₈O₃ |
MDL Number:
MFCD12827902 CAS Number: 1215661-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...