2-(4-Bromophenoxy)-6-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

083800-1G 083800-10G
102840-764EA 252.31 USD
102840-764 102840-766
2-(4-Bromophenoxy)-6-(trifluoromethyl)pyridine ≥97%
2-(4-Bromophenoxy)-6-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₂H₇BrF₃NO
MDL Number: MFCD10703540
CAS Number: 1086376-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR