2-(4-Bromophenoxy)-5-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

083799-1G 083799-10G
102840-760EA 383.08 USD
102840-760 102840-762
2-(4-Bromophenoxy)-5-(trifluoromethyl)pyridine ≥97%
2-(4-Bromophenoxy)-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₂H₇BrF₃NO
MDL Number: MFCD00204096
CAS Number: 137736-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR