2-(4-Bromophenoxy)-3-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

083798-1G 083798-10G
102840-756EA 383.08 USD
102840-756 102840-758
2-(4-Bromophenoxy)-3-(trifluoromethyl)pyridine ≥97%
2-(4-Bromophenoxy)-3-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₂H₇BrF₃NO
MDL Number: MFCD10703539
CAS Number: 1019441-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR