4-(3-Bromophenoxy)-2-(trifluoromethyl)pyrimidine ≥97%
Supplier: Matrix Scientific
083791-1G
083791-10G
102840-738EA
383.08
USD
102840-738
102840-740
4-(3-Bromophenoxy)-2-(trifluoromethyl)pyrimidine ≥97%
4-(3-Bromophenoxy)-2-(trifluoromethyl)pyrimidine
Hazard: Irritant
Formula:
C₁₁H₆BrF₃N₂O |
MDL Number:
MFCD09864946 CAS Number: 1086376-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...