Ethyl-4-chloro-1-benzothiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

083528-1G 083528-10G
102840-012EA 353.85 USD
102840-012 102840-014
Ethyl-4-chloro-1-benzothiophene-2-carboxylate ≥97%
Ethyl-4-chloro-1-benzothiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₉ClO₂S
MW: 240.71 g/mol
MDL Number: MFCD11100113
CAS Number: 1207537-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR