Ethyl-6-(trifluoromethyl)-1-benzothiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

083525-1G 083525-10G
102840-004EA 353.85 USD
102840-004 102840-006
Ethyl-6-(trifluoromethyl)-1-benzothiophene-2-carboxylate ≥97%
Ethyl-6-(trifluoromethyl)-1-benzothiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₉F₃O₂S
MW: 274.26 g/mol
MDL Number: MFCD11100110
CAS Number: 1132650-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR