Diethyl (4-chloro-2-fluorophenyl)malonate ≥97%

Supplier: Matrix Scientific

Synonyms: Diethyl 2-(4-chloro-2-fluorophenyl)malonate

083371-1G 083371-10G
102839-598EA 292.31 USD
102839-598 102839-600
Diethyl (4-chloro-2-fluorophenyl)malonate ≥97%
Diethyl (4-chloro-2-fluorophenyl)malonate

Hazard: Irritant

Formula: C₁₃H₁₄ClFO₄
MW: 288.71 g/mol
MDL Number: MFCD01570544
CAS Number: 680211-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR