Methyl-3-[(3-chloro-2-formylphenyl)thio]propanoate ≥97%

Supplier: Matrix Scientific

083352-1G 083352-10G
102839-534EA 383.08 USD
102839-534 102839-536
Methyl-3-[(3-chloro-2-formylphenyl)thio]propanoate ≥97%
Methyl-3-[(3-chloro-2-formylphenyl)thio]propanoate

Hazard: Irritant

Formula: C₁₁H₁₁ClO₃S
MW: 258.73 g/mol
MDL Number: MFCD11100102
CAS Number: 1215582-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR