Ethyl-3-({[(2-chloroethyl)amino]carbonyl}amino)benzoate ≥97%

Supplier: Matrix Scientific

083351-1G 083351-10G
102839-530EA 226.15 USD
102839-530 102839-532
Ethyl-3-({[(2-chloroethyl)amino]carbonyl}amino)benzoate ≥97%
Ethyl-3-({[(2-chloroethyl)amino]carbonyl}amino)benzoate

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₃
MW: 270.72 g/mol
MDL Number: MFCD09836176
CAS Number: 803729-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR