1-((4-(tert-Butyl)phenyl)sulfonyl)-2-(hydroxyimino)eth-2-ylamine ≥95%

Supplier: Matrix Scientific

084590-1G 084590-10G
102842-828EA 467.69 USD
102842-828 102842-830
1-((4-(tert-Butyl)phenyl)sulfonyl)-2-(hydroxyimino)eth-2-ylamine ≥95%
1-((4-(tert-Butyl)phenyl)sulfonyl)-2-(hydroxyimino)eth-2-ylamine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₃S
MW: 270.35 g/mol
MDL Number: MFCD03410410
CAS Number: 884504-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR