4-(4-Methylphenoxy)benzenesulfonyl chloride ≥95%
Supplier: Matrix Scientific
084404-1G
084404-10G
102842-320EA
707.69
USD
102842-320
102842-322
4-(4-Methylphenoxy)benzenesulfonyl chloride ≥95%
4-(4-Methylphenoxy)benzenesulfonyl chloride
Hazard: Irritant
Formula:
C₁₃H₁₁ClO₃S |
MDL Number:
MFCD01631869 CAS Number: 192329-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...