4-Bromo-1,1,3,3,6-pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-ylamine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Amino-4-bromo-7-nitro-1,1,3,3,6-pentamethylindane

084145-1G 084145-10G
102841-624EA 270.77 USD
102841-624 102841-626
4-Bromo-1,1,3,3,6-pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-ylamine ≥97%
4-Bromo-1,1,3,3,6-pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-ylamine

Hazard: Irritant

Formula: C₁₄H₁₉BrN₂O₂
MW: 327.23 g/mol
MDL Number: MFCD11100123
CAS Number: 1210378-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR