3-{4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl}propanoic acid ≥97%
Supplier: Matrix Scientific
082766-1G
082766-10G
102838-076EA
298.46
USD
102838-076
102838-078
3-{4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl}propanoic acid ≥97%
3-{4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl}propanoic acid
Hazard: Irritant
Formula:
C₁₃H₁₆F₃N₃O₂ |
MDL Number:
MFCD11035862 CAS Number: 1031929-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...