2-[5-(Trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082585-1G 082585-10G
102837-662EA 566.15 USD
102837-662 102837-664
2-[5-(Trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid ≥97%
2-[5-(Trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₈H₄F₃N₃O₂S
MDL Number: MFCD09836179
CAS Number: 1209904-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR