Methyl-N-benzoyl-2-methylalaninate

Supplier: Matrix Scientific

081052-500MG 081052-1G
102672-552EA 258.46 USD
102672-552 102672-554
Methyl-N-benzoyl-2-methylalaninate
Methyl-N-benzoyl-2-methylalaninate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD01413846
CAS Number: 65563-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR