4-Ethyl-3-methylpiperazin-2-one hydrochloride

Supplier: Matrix Scientific

081044-500MG 081044-1G
102672-514EA 516.92 USD
102672-514 102672-516
4-Ethyl-3-methylpiperazin-2-one hydrochloride
4-Ethyl-3-methylpiperazin-2-one hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O
MDL Number: MFCD22422047
CAS Number: 1417566-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR