Ethyl-2-bromo-4-methyl-1H-imidazole-5-carboxylate hydrochloride
Supplier: Matrix Scientific
081039-500MG
081039-1G
102672-496EA
516.92
USD
102672-496
102672-498
Ethyl-2-bromo-4-methyl-1H-imidazole-5-carboxylate hydrochloride
Ethyl-2-bromo-4-methyl-1H-imidazole-5-carboxylate hydrochloride
Hazard: Irritant
Formula:
C₇H₁₀BrClN₂O₂ |
MDL Number:
MFCD22422045 CAS Number: 1417569-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...