1-(2-Chloroethyl)-3,5-dimethyl-1H-1,2,4-triazole hydrochloride

Supplier: Matrix Scientific

081038-500MG 081038-1G
102672-492EA 516.92 USD
102672-492 102672-494
1-(2-Chloroethyl)-3,5-dimethyl-1H-1,2,4-triazole hydrochloride
1-(2-Chloroethyl)-3,5-dimethyl-1H-1,2,4-triazole hydrochloride

Hazard: Irritant

Formula: C₆H₁₁Cl₂N₃
MDL Number: MFCD22056462
CAS Number: 53946-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR