1-Amino-3-(dimethylamino)propan-2-ol hydrochloride
Supplier: Matrix Scientific
081032-1G
081032-5G
102672-470EA
646.15
USD
102672-470
102672-472
1-Amino-3-(dimethylamino)propan-2-ol hydrochloride
1-Amino-3-(dimethylamino)propan-2-ol hydrochloride
Hazard: Irritant
Formula:
C₅H₁₅ClN₂O |
MDL Number:
MFCD22422044 CAS Number: 1417567-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...