1-(1-Methylpiperidin-4-yl)azepane

Supplier: Matrix Scientific

081029-500MG 081029-1G 081029-5G
102672-450EA 161.54 USD
102672-450 102672-452 102672-454
1-(1-Methylpiperidin-4-yl)azepane
1-(1-Methylpiperidin-4-yl)azepane

Hazard: Irritant

Formula: C₁₂H₂₄N₂
MDL Number: MFCD22422043
CAS Number: 1394306-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR