4-Bromo-3-(methoxymethyl)-5-methyl-1H-pyrazole

Supplier: Matrix Scientific

081026-500MG 081026-1G
102672-438EA 516.92 USD
102672-438 102672-440
4-Bromo-3-(methoxymethyl)-5-methyl-1H-pyrazole
4-Bromo-3-(methoxymethyl)-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₆H₉BrN₂O
MW: 205.06 g/mol
MDL Number: MFCD20502768
CAS Number: 1287752-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR