3-(Methoxymethyl)-5-methyl-1H-pyrazole

Supplier: Matrix Scientific

081025-500MG 081025-1G
102672-434EA 646.15 USD
102672-434 102672-436
3-(Methoxymethyl)-5-methyl-1H-pyrazole
3-(Methoxymethyl)-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₆H₁₀N₂O
MDL Number: MFCD22422041
CAS Number: 37599-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR