[(6,6-Dimethylbicyclo[3.1.1]hept-2-yl)methyl]amine

Supplier: Matrix Scientific

081013-500MG 081013-1G 081013-5G
102672-378EA 516.92 USD
102672-378 102672-380 102672-382
[(6,6-Dimethylbicyclo[3.1.1]hept-2-yl)methyl]amine
[(6,6-Dimethylbicyclo[3.1.1]hept-2-yl)methyl]amine

Hazard: Irritant

Formula: C₁₀H₁₉N
MDL Number: MFCD07809522
CAS Number: 74837-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR