N-[(2E)-2-Amino-2-(hydroxyimino)ethyl]acetamide
Supplier: Matrix Scientific
081009-500MG
081009-1G
102672-356EA
646.15
USD
102672-356
102672-358
N-[(2E)-2-Amino-2-(hydroxyimino)ethyl]acetamide
N-[(2E)-2-Amino-2-(hydroxyimino)ethyl]acetamide
Hazard: Irritant
Formula:
C₄H₉N₃O₂ |
MDL Number:
MFCD22422039 CAS Number: 390817-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...