4-(Chloromethyl)-N,N-dimethylbenzamide

Supplier: Matrix Scientific

081003-500MG 081003-1G 081003-5G
102672-326EA 710.77 USD
102672-326 102672-328 102672-330
4-(Chloromethyl)-N,N-dimethylbenzamide
4-(Chloromethyl)-N,N-dimethylbenzamide

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
MDL Number: MFCD11204892
CAS Number: 121083-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR