5-Cyclopropyl-1,2-dihydro-3H-1,2,4-triazole-3-thione

Supplier: Matrix Scientific

081002-500MG 081002-1G
102672-322EA 516.92 USD
102672-322 102672-324
5-Cyclopropyl-1,2-dihydro-3H-1,2,4-triazole-3-thione
5-Cyclopropyl-1,2-dihydro-3H-1,2,4-triazole-3-thione

Hazard: Irritant

Formula: C₅H₇N₃S
MDL Number: MFCD22422036
CAS Number: 75570-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR