3-Chloro-5-cyclopropyl-1,2,4-oxadiazole

Supplier: Matrix Scientific

080993-500MG 080993-1G
102672-276EA 646.15 USD
102672-276 102672-278
3-Chloro-5-cyclopropyl-1,2,4-oxadiazole
3-Chloro-5-cyclopropyl-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₅H₅ClN₂O
MDL Number: MFCD22422033
CAS Number: 1393567-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR