3-(Chloromethyl)-5-(2-phenylethyl)-1,2,4-oxadiazole
Supplier: Matrix Scientific
080985-500MG
080985-1G
102672-234EA
387.69
USD
102672-234
102672-236
3-(Chloromethyl)-5-(2-phenylethyl)-1,2,4-oxadiazole
3-(Chloromethyl)-5-(2-phenylethyl)-1,2,4-oxadiazole
Hazard: Irritant
Formula:
C₁₁H₁₁ClN₂O |
MDL Number:
MFCD10695126 CAS Number: 1152498-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...