4-(3-Amino-1H-1,2,4-triazol-5-yl)-1-methylpyrrolidin-2-one

Supplier: Matrix Scientific

080979-500MG 080979-1G 080979-5G
102672-202EA 516.92 USD
102672-202 102672-204 102672-206
4-(3-Amino-1H-1,2,4-triazol-5-yl)-1-methylpyrrolidin-2-one
4-(3-Amino-1H-1,2,4-triazol-5-yl)-1-methylpyrrolidin-2-one

Hazard: Irritant

Formula: C₇H₁₁N₅O
MDL Number: MFCD20502374
CAS Number: 1351393-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR