2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptylbenzenesulfonate

Supplier: Matrix Scientific

080976-500MG 080976-1G 080976-5G
102672-184EA 161.54 USD
102672-184 102672-186 102672-188
2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptylbenzenesulfonate
2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptylbenzenesulfonate

Hazard: Irritant

Formula: C₁₃H₈F₁₂O₃S
MDL Number: MFCD00344899
CAS Number: 85459-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR