5-Oxa-7-azaspiro[2.5]octan-6-one
Supplier: Matrix Scientific
080965-500MG
080965-1G
080965-5G
102672-126EA
646.15
USD
102672-126
102672-128
102672-130
5-Oxa-7-azaspiro[2.5]octan-6-one
5-Oxa-7-azaspiro[2.5]octan-6-one
Hazard: Irritant
Formula:
C₆H₉NO₂ |
MDL Number:
MFCD22422022 CAS Number: 1394306-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...