4-Iodo-1H-pyrazol-5-amine

Supplier: Matrix Scientific

Synonyms: 3-Amino-4-iodo-1H-pyrazole, 4-Iodo-1H-pyrazol-3-amine, 4-Iodo-3-aminopyrazole

080961-500MG 080961-1G 080961-5G
102672-102EA 338.46 USD
102672-102 102672-104 102672-106
4-Iodo-1H-pyrazol-5-amine
4-Iodo-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₃H₄IN₃
MW: 208.99 g/mol
MDL Number: MFCD22422021
CAS Number: 81542-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR