2-Amino-7-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid
Supplier: Matrix Scientific
080959-500MG
080959-1G
102672-092EA
387.69
USD
102672-092
102672-094
2-Amino-7-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid
2-Amino-7-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid
Hazard: Irritant
Formula:
C₇H₇N₅O₂ |
MDL Number:
MFCD22422020 CAS Number: 1394306-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...