1-(2-Chloroethyl)-1H-benzimidazole hydrochloride

Supplier: Matrix Scientific

080955-500MG 080955-1G
102672-072EA 387.69 USD
102672-072 102672-074
1-(2-Chloroethyl)-1H-benzimidazole hydrochloride
1-(2-Chloroethyl)-1H-benzimidazole hydrochloride

Hazard: Irritant

Formula: C₉H₁₀Cl₂N₂
MDL Number: MFCD22422018
CAS Number: 26706-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR