4-Ethylpiperazin-2-one hydrochloride

Supplier: Matrix Scientific

080953-500MG 080953-1G
102672-066EA 292.31 USD
102672-066 102672-068
4-Ethylpiperazin-2-one hydrochloride
4-Ethylpiperazin-2-one hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O
MDL Number: MFCD22422017
CAS Number: 65464-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR