1-(Trifluoroacetyl)piperidine-3-carboxylic acid
Supplier: Matrix Scientific
080949-500MG
080949-1G
080949-5G
102672-046EA
387.69
USD
102672-046
102672-048
102672-050
1-(Trifluoroacetyl)piperidine-3-carboxylic acid
1-(Trifluoroacetyl)piperidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₈H₁₀F₃NO₃ |
MDL Number:
MFCD09908273 CAS Number: 1099017-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...