N-(2-Phenylethyl)but-3-en-1-amine
Supplier: Matrix Scientific
080945-500MG
080945-1G
102672-028EA
338.46
USD
102672-028
102672-030
N-(2-Phenylethyl)but-3-en-1-amine
N-(2-Phenylethyl)but-3-en-1-amine
Hazard: Irritant
Formula:
C₁₂H₁₇N |
MDL Number:
MFCD20289163 CAS Number: 867324-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...