4'-(Propylsulfamoyl)acetanilide

Supplier: Matrix Scientific

080914-500MG 080914-1G 080914-5G
102671-870EA 161.54 USD
102671-870 102671-872 102671-874
4'-(Propylsulfamoyl)acetanilide
4'-(Propylsulphamoyl)acetanilide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃S
MDL Number: MFCD01213956
CAS Number: 349398-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR