N-(3-Hydroxyphenyl)-5-oxo-1-phenylpyrrolidine-3-carboxamide
Supplier: Matrix Scientific
080911-500MG
080911-1G
102671-856EA
193.85
USD
102671-856
102671-858
N-(3-Hydroxyphenyl)-5-oxo-1-phenylpyrrolidine-3-carboxamide
N-(3-Hydroxyphenyl)-5-oxo-1-phenylpyrrolidine-3-carboxamide
Hazard: Irritant
Formula:
C₁₇H₁₆N₂O₃ |
MDL Number:
MFCD02647341 CAS Number: 352330-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...