N-(5-Chloro-2-hydroxyphenyl)-5-oxo-1-phenylpyrrolidine-3-carboxamide

Supplier: Matrix Scientific

080910-500MG 080910-1G
102671-852EA 193.85 USD
102671-852 102671-854
N-(5-Chloro-2-hydroxyphenyl)-5-oxo-1-phenylpyrrolidine-3-carboxamide
N-(5-Chloro-2-hydroxyphenyl)-5-oxo-1-phenylpyrrolidine-3-carboxamide

Hazard: Irritant

Formula: C₁₇H₁₅ClN₂O₃
MDL Number: MFCD02647340
CAS Number: 361369-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR