3-{3-[4-(Benzyloxy)-3-methoxyphenyl]-1,2,4-oxadiazol-5-yl}propanoic acid

Supplier: Matrix Scientific

080907-500MG 080907-1G
102671-844EA 258.46 USD
102671-844 102671-846
3-{3-[4-(Benzyloxy)-3-methoxyphenyl]-1,2,4-oxadiazol-5-yl}propanoic acid
3-{3-[4-(Benzyloxy)-3-methoxyphenyl]-1,2,4-oxadiazol-5-yl}propanoic acid

Hazard: Irritant

Formula: C₁₉H₁₈N₂O₅
MDL Number: MFCD08133361
CAS Number: 889946-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR