2-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]ethyl chloroacetate

Supplier: Matrix Scientific

080904-500MG 080904-1G 080904-5G
102671-830EA 193.85 USD
102671-830 102671-832 102671-834
2-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]ethyl chloroacetate
2-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]ethyl chloroacetate

Hazard: Irritant

Formula: C₁₁H₁₁ClN₂O₄S
MDL Number: MFCD03215159
CAS Number: 591212-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR