1-[4-(1,1-Dioxido-1,2-benzisothiazol-3-yl)piperazin-1-yl]-3-methoxypropan-2-olhydrochloride

Supplier: Matrix Scientific

080902-500MG 080902-1G
102671-822EA 193.85 USD
102671-822 102671-824
1-[4-(1,1-Dioxido-1,2-benzisothiazol-3-yl)piperazin-1-yl]-3-methoxypropan-2-olhydrochloride
1-[4-(1,1-Dioxido-1,2-benzisothiazol-3-yl)piperazin-1-yl]-3-methoxypropan-2-olhydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClN₃O₄S
MDL Number: MFCD03719950
CAS Number: 1052150-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR