(6-Bromo-2-oxo-1,3-benzoxazol-3(2H)-yl)acetic acid

Supplier: Matrix Scientific

080896-500MG 080896-1G 080896-5G
102671-804EA 387.69 USD
102671-804 102671-806 102671-808
(6-Bromo-2-oxo-1,3-benzoxazol-3(2H)-yl)acetic acid
(6-Bromo-2-oxo-1,3-benzoxazol-3(2H)-yl)acetic acid

Hazard: Irritant

Formula: C₉H₆BrNO₄
MDL Number: MFCD00700893
CAS Number: 100219-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR